Learn More
Cayman Chemical B71CD80 LNg Isoform Extrac 1mg
SDP
Supplier:  Cayman Chemical 320131 MG
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 25.4 kDa
Catalog No. 50-294-7106