Learn More
ACROBiosystems Recombinant Protein;Human B7-1 / CD80 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;B71,CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7
Supplier:  ACROBiosystems B71H52A41MG
Encompass_Preferred
Recombinant Protein;Human B7-1 / CD80 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human B7-1, mouse IgG2a Fc Tag, low endotoxin (B71-H52A4) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # P33681-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 50.9 kDa. The protein migrates as 65-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CTLA-4, His Tag (Cat. No. CT4-H5229) at 1 μg/mL (100 μL/well) can bind Human B7-1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Human B7-1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) at 5 μg/mL (100 μL/well) can bind Human PD-L1, Fc Tag (Cat. No. PD1-H5258) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-165-8162