Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag;HEK293;1MG;CD6,CD86,CD-86,B7-2,B70,B-70,CD28LG2,LAB72,LAB-72,MGC34413
Catalog No. 501659129
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-2, Fc Tag (Cat. No. CD6-C5254) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-165-9129 Supplier ACROBiosystems Supplier No. CD6C52541MG
Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-2, Fc Tag (Cat. No. CD6-C5254) with a linear range of 0.039-1.25 μg/mL (QC tested).