Learn More
ACROBiosystems Recombinant Protein;Mouse B7-2 / CD86 Protein, Fc Tag;HEK293;1MG;CD6,CD86,CD-86,B7-2,B70,B-70,CD28LG2,LAB72,LAB-72,MGC34413
Catalog No. 501658754
Encompass_Preferred
Recombinant Protein;Mouse B7-2 / CD86 Protein, Fc Tag;1MG;Mouse B7-2, Fc Tag (CD6-M5251) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession # NP_062261.3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-2, Fc Tag (Cat. No. CD6-M5251) at 5 μg/mL (100 μL/well) can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-8754 Supplier ACROBiosystems Supplier No. CD6M52511MG
Recombinant Protein;Mouse B7-2 / CD86 Protein, Fc Tag;1MG;Mouse B7-2, Fc Tag (CD6-M5251) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession # NP_062261.3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-2, Fc Tag (Cat. No. CD6-M5251) at 5 μg/mL (100 μL/well) can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.4-13 ng/mL (QC tested).