Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein (HPLC-verified);HEK293;100UG;CD6,CD86,CD-86,B7-2,B70,B-70,CD28LG2,LAB72,LAB-72,MGC34413
Catalog No. 501659130
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein (HPLC-verified);100UG;Cynomolgus / Rhesus macaque B7-2, His Tag (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa. The protein migrates as 50-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9130 Supplier ACROBiosystems Supplier No. CD6C52H5100UG
Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein (HPLC-verified);100UG;Cynomolgus / Rhesus macaque B7-2, His Tag (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa. The protein migrates as 50-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.