Learn More
Cayman Chemical B7-CD28 Interaction Inhibitor, Purity: ≥95%, CAS: 635324-72-0, 1 mg
SDP
Catalog No. 502943948
Encompass_Preferred
An inhibitor of the B7-1-CD28 interaction (IC50 = 50 nM)
Catalog No. 50-294-3948 Supplier Cayman Chemical Supplier No. 423981 MG
An inhibitor of the B7-1-CD28 interaction (IC50 = 50 nM)