Learn More
ACROBiosystems Recombinant Protein;Human B7-H2 / ICOSLG Protein;HEK293;200UG;B72,ICOSLG,B7-H2,B7H2,B7RP-1,B7RP1,CD275,GL50,ICOS-L,ICOSL,LICOS,ICOS ligand
Supplier: ACROBiosystems B72H5221200UG
Recombinant Protein;Human B7-H2 / ICOSLG Protein;200UG;Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession # NP_056074).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.5 kDa. The protein migrates as 35-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, His Tag (Cat. No. B72-H5221) at 1 μg/mL (100 µl/well),can bind Human ICOS, Fc Tag (Cat. No. ICS-H5250) with a linear of 0.07-2.3 ng/mL (QC tested).