Learn More
Sino Biological B7-H3 / CD276 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 80380T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat B7-H3 / CD276 (Catalog#80380-R08H; Q7TPB4; Met1-Phe244). B7-H3 / CD276 specific IgG was purified by Rat B7-H3 / CD276 affinity chromatography.
Catalog No. 50-224-8347