Learn More
Cayman Chemical B7H3CD276 EXrAClular Dom 100ug
SDP
Catalog No. 502949192
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-tagged B7-H3 expressed in HEK293 cells • Amino acids: 27-461 • MW: 73.4 kDa
Catalog No. 50-294-9192 Supplier Cayman Chemical Supplier No. 31822100 UG
Source: Recombinant C-terminal human IgG1 Fc-tagged B7-H3 expressed in HEK293 cells • Amino acids: 27-461 • MW: 73.4 kDa