Learn More
Cayman Chemical B7H3CD276CTerm RabbIt M 100uL
SDP
Catalog No. 502946531
Encompass_Preferred
A quaternary ammonium-containing cationic surfactant with diverse biological activities an inhibitor of F. tularensis AcpB (Ki 0.59 UM) and C. albicans V-ATPase (EC50 69 UM) inhibits osteoclastogenesis RANKL-induced osteoclastogenesis in BMDMs inhibits bone resorption by mature osteoclasts at 100 and 200 nM prevents LPS-induced bone loss in a calvarial osteolytic mouse model at 5 mg/kg per day induces mortality in C. oncophora O. ostertagi H. contortus and T. circumcincta (EC50s 4.5 10.8 5.1 and 6.7 UM respectively) but is cytotoxic to HEK293 and RAW 264.7 cells (CC50s 9.2 and 5.7 UM respectively)
Catalog No. 50-294-6531 Supplier Cayman Chemical Supplier No. 32272100 UL
A quaternary ammonium-containing cationic surfactant with diverse biological activities an inhibitor of F. tularensis AcpB (Ki 0.59 UM) and C. albicans V-ATPase (EC50 69 UM) inhibits osteoclastogenesis RANKL-induced osteoclastogenesis in BMDMs inhibits bone resorption by mature osteoclasts at 100 and 200 nM prevents LPS-induced bone loss in a calvarial osteolytic mouse model at 5 mg/kg per day induces mortality in C. oncophora O. ostertagi H. contortus and T. circumcincta (EC50s 4.5 10.8 5.1 and 6.7 UM respectively) but is cytotoxic to HEK293 and RAW 264.7 cells (CC50s 9.2 and 5.7 UM respectively)