Learn More
ACROBiosystems Recombinant Protein;Human B7-H3 / CD276 Protein, Fc Tag;HEK293;100UG;B73,B7-H3,CD276,B7 homolog 3,B7H3
Catalog No. 501659034
Encompass_Preferred
Recombinant Protein;Human B7-H3 / CD276 Protein, Fc Tag;100UG;Human B7-H3, Fc Tag (B73-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9034 Supplier ACROBiosystems Supplier No. B73H5253100UG
Recombinant Protein;Human B7-H3 / CD276 Protein, Fc Tag;100UG;Human B7-H3, Fc Tag (B73-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.