Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human B7-H3 / CD276 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;B73,B7-H3,CD276,B7 homolog 3,B7H3
Supplier:  ACROBiosystems B73H82F525UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human B7-H3 / CD276 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human B7-H3, Fc Tag, Avi Tag (B73-H82F5) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.6 kDa. The protein migrates as 63-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7921