Learn More
ACROBiosystems Recombinant Protein;Human B7-H3 / CD276 Protein;HEK293;1MG;B73,B7-H3,CD276,B7 homolog 3,B7H3
Catalog No. 501659037
Encompass_Preferred
Recombinant Protein;Human B7-H3 / CD276 Protein;1MG;Human B7-H3, His Tag (B73-H52E2) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.2 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9037 Supplier ACROBiosystems Supplier No. B73H52E21MG
Recombinant Protein;Human B7-H3 / CD276 Protein;1MG;Human B7-H3, His Tag (B73-H52E2) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.2 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.