Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human B7-H4, ultra sensitivity (primary amine labeling);HEK293;200UG;B74,B7-H4,VTCN1,B7H4,B7S1,B7h.5,VTCN-1,B7-S1
Supplier:  ACROBiosystems B74H8222200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human B7-H4, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human B7-H4 (B74-H8222) is expressed from human HEK293 cells. It contains AA Phe 29 - Ala 258 (Accession # NP_078902). It is the biotinylated form of Human B7-H4 protein, His Tag (Cat. No. B74-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.1 kDa. The protein migrates as 43-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human B7-H4 MAb at 2 g/mL (100 L/well) can bind Biotinylated Human B7-H4, His Tag (Cat. No. B74-H8222) with a linear range of 1-8 ng/mL (QC tested).
Catalog No. 50-165-9183