Learn More
ACROBiosystems Recombinant Protein;Mouse B7-H5 / Gi24 / VISTA Protein;HEK293;100UG;B75,B7-H5,C10orf54,Sisp-1,SISP1,Gi24,VISTA,B7H5
Supplier:  ACROBiosystems B75M52H7100UG
Encompass_Preferred
Recombinant Protein;Mouse B7-H5 / Gi24 / VISTA Protein;100UG;Mouse B7-H5, His Tag (B75-M52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 191 (Accession # NP_001153044.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.7kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8420