Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque B7-H5 / Gi24 / VISTA Protein;HEK293;1MG;B75,B7-H5,C10orf54,Sisp-1,SISP1,Gi24,VISTA,B7H5
Catalog No. 501658424
Encompass_Preferred
Recombinant Protein;Rhesus macaque B7-H5 / Gi24 / VISTA Protein;1MG;Rhesus macaque B7-H5, His Tag (B75-C5220) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Thr 192 (Accession # NP_001177807.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8424 Supplier ACROBiosystems Supplier No. B75C52201MG
Recombinant Protein;Rhesus macaque B7-H5 / Gi24 / VISTA Protein;1MG;Rhesus macaque B7-H5, His Tag (B75-C5220) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Thr 192 (Accession # NP_001177807.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.