Learn More
ACROBiosystems Recombinant Protein;Human B7-H5 / Gi24 / VISTA Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;B75,B7-H5,C10orf54,Sisp-1,SISP1,Gi24,VISTA,B7H5
Catalog No. 501658093
Encompass_Preferred
Recombinant Protein;Human B7-H5 / Gi24 / VISTA Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (B75-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8093 Supplier ACROBiosystems Supplier No. B75H52581MG
Recombinant Protein;Human B7-H5 / Gi24 / VISTA Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (B75-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.