Learn More
ACROBiosystems Recombinant Protein;Cynomolgus B7-H6 / NCR3LG1 Protein;HEK293;100UG;B76,B7-H6,NCR3LG1,B7 Homolog 6,B7H6
Supplier:  ACROBiosystems B76C52HA100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus B7-H6 / NCR3LG1 Protein;100UG;Cynomolgus B7-H6 (B76-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Asp 259 (Accession # XP_005578557.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.4 kDa. The protein migrates as 38-50 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NKp30, Fc Tag (Cat. No. NC3-H5259) at 5 μg/mL (100 μL/well) can bind Cynomolgus B7-H6 (Cat. No. B76-C52Ha) with a linear range of 0.625-5 μg/mL (QC tested).
Catalog No. 50-165-7704