Learn More
ACROBiosystems Recombinant Protein;Human B7-H6 / NCR3LG1 Protein;HEK293;100UG;B76,B7-H6,NCR3LG1,B7 Homolog 6,B7H6
Supplier:  ACROBiosystems B76H52H8100UG
Encompass_Preferred
Recombinant Protein;Human B7-H6 / NCR3LG1 Protein;100UG;Human B7-H6 (B76-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Accession # Q68D85-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.6 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NKp30, Fc Tag (Cat. No. NC3-H5259) at 10 μg/mL (100 μL/well) can bind Human B7-H6 (Cat. No. B76-H52H8) with a linear range of 0.078-0.313 μg/mL (QC tested).
Catalog No. 50-165-7682