Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human B7-H6 / NCR3LG1 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;B76,B7-H6,NCR3LG1,B7 Homolog 6,B7H6
Supplier:  ACROBiosystems B76H82E525UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human B7-H6 / NCR3LG1 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human B7-H6, His Tag, Avi Tag (B76-H82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Accession # Q68D85-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 30.4 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8271