Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human B7-H7 / HHLA2 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;B77,B7-H7,HHLA2,B7 Homolog 7,B7H7
Catalog No. 501658078
Encompass_Preferred
Recombinant Protein;Biotinylated Human B7-H7 / HHLA2 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human B7-H7, Fc Tag, Avi Tag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 65.9 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, Fc Tag (Cat. No. CDH-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-H7, Fc Tag, Avi Tag (Cat. No. B77-H82F5) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-165-8078 Supplier ACROBiosystems Supplier No. B77H82F5200UG
Recombinant Protein;Biotinylated Human B7-H7 / HHLA2 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human B7-H7, Fc Tag, Avi Tag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 65.9 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, Fc Tag (Cat. No. CDH-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-H7, Fc Tag, Avi Tag (Cat. No. B77-H82F5) with a linear range of 5-78 ng/mL (QC tested).