Learn More
Medchemexpress LLC B8R 20-27 | 848642-31-9 | 99.38% | 960.04 | 1 MG
SDP
Catalog No. 5000396920
Encompass_Preferred
B8R 20-27 is a biologically active peptide. It is an 8-amino acid fragment (amino acids 20 to 27) of B8R, a vaccinia virus (VV) gene that encodes a secreted protein related to the gamma interferon receptor. B8R binding to IFN-g neutralizes its antiviral activity.
  • Biologically active peptide
  • Fragment of B8R vaccinia virus gene
  • Related to gamma interferon receptor
  • Neutralizes IFN-g antiviral activity
Catalog No. 50-003-96920 Supplier Medchemexpress LLC Supplier No. HYP53641MG
B8R 20-27 is a biologically active peptide. It is an 8-amino acid fragment (amino acids 20 to 27) of B8R, a vaccinia virus (VV) gene that encodes a secreted protein related to the gamma interferon receptor. B8R binding to IFN-g neutralizes its antiviral activity.
  • Biologically active peptide
  • Fragment of B8R vaccinia virus gene
  • Related to gamma interferon receptor
  • Neutralizes IFN-g antiviral activity