Learn More
CPC Scientific H-DArg-Arg-Pro-Hyp-Gly-Thi-Ser-DIgl-Oic-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108859
Encompass
ONE-LETTER SEQUENCE: rRP-Hyp-G-Thi-S-DIgl-Oic-R MOLECULAR FORMULA: C60H90N19O13S1MOLECULAR WEIGHT:1317.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [175413-73-7], RESEARCH AREA: Cardiovascular
Catalog No. 50-210-8859 Supplier CPC Scientific Supplier No. BRAK008A
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ONE-LETTER SEQUENCE: rRP-Hyp-G-Thi-S-DIgl-Oic-R MOLECULAR FORMULA: C60H90N19O13S1MOLECULAR WEIGHT:1317.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [175413-73-7], RESEARCH AREA: Cardiovascular