Learn More
CPC Scientific H-DArg-Arg-Pro-Hyp-Gly-Igl-Ser-DIgl-Oic-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108861
Encompass
ONE-LETTER SEQUENCE: rRP-Hyp-G-Igl-S-DIgl-Oic-R MOLECULAR FORMULA: C64H94N19O13MOLECULAR WEIGHT:1338.58STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [180981-09-3], SYNONYMS: B-9430, (D-Arg,Hyp,Igl,D-Igl,Oic)-BradykininRESEARCH AREA: Cardiovascular
Catalog No. 50-210-8861 Supplier CPC Scientific Supplier No. BRAK009A
ONE-LETTER SEQUENCE: rRP-Hyp-G-Igl-S-DIgl-Oic-R MOLECULAR FORMULA: C64H94N19O13MOLECULAR WEIGHT:1338.58STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [180981-09-3], SYNONYMS: B-9430, (D-Arg,Hyp,Igl,D-Igl,Oic)-BradykininRESEARCH AREA: Cardiovascular