Learn More
Medchemexpress LLC BA 1 TFA | 99.4% | 1247.32 | 5 MG
SDP
Catalog No. 5000423682
Encompass_Preferred
BA 1 TFA is a potent agonist for the bombesin (BB) family of receptors. It binds with high affinity to Bombesin receptor subtype-3 (BRS3), gastrin releasing peptide receptor (GRPR), and neuromedin B receptor (NMBR) with IC50s of 6, 0.4, and 2.5 nM respectively. BA1 (0.1 μM) significantly increases FAK tyrosine phosphorylation in BRS-3 transfected NCI-H1299 cells.
  • Potent agonist for bombesin (BB) family of receptors
  • Binds with high affinity to BRS3 (IC50 6 nM)
  • Binds with high affinity to GRPR (IC50 0.4 nM)
  • Binds with high affinity to NMBR (IC50 2.5 nM)
  • Significantly increases FAK tyrosine phosphorylation in BRS-3 transfected NCI-H1299 cells
Catalog No. 50-004-23682 Supplier Medchemexpress LLC Supplier No. HYP1423A5MG
BA 1 TFA is a potent agonist for the bombesin (BB) family of receptors. It binds with high affinity to Bombesin receptor subtype-3 (BRS3), gastrin releasing peptide receptor (GRPR), and neuromedin B receptor (NMBR) with IC50s of 6, 0.4, and 2.5 nM respectively. BA1 (0.1 μM) significantly increases FAK tyrosine phosphorylation in BRS-3 transfected NCI-H1299 cells.
  • Potent agonist for bombesin (BB) family of receptors
  • Binds with high affinity to BRS3 (IC50 6 nM)
  • Binds with high affinity to GRPR (IC50 0.4 nM)
  • Binds with high affinity to NMBR (IC50 2.5 nM)
  • Significantly increases FAK tyrosine phosphorylation in BRS-3 transfected NCI-H1299 cells