Learn More
Medchemexpress LLC BA-53038B 100mg | 2306195-65-1 | 100 MG
SDP
Catalog No. 5000229622
Encompass_Preferred
BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator used in research to modulate capsid assembly and assess antiviral activity (reported EC50 3.32 μM). It is supplied as a high-purity solid and as DMSO stock solutions for in vitro studies.
  • Acts as an HBV core protein allosteric modulator targeting the HAP pocket.
  • Demonstrated antiviral activity (EC50 3.32 μM) in cell-based assays.
  • High purity (98.73%) and white to off-white solid appearance.
  • Chemical formula C14H16ClNO; molecular weight 249.74 g·mol⁻¹.
  • Available as solid quantities (up to 100 mg) and as a 10 mM DMSO solution.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.
Catalog No. 50-002-29622 Supplier Medchemexpress LLC Supplier No. HY114314100MG
BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator used in research to modulate capsid assembly and assess antiviral activity (reported EC50 3.32 μM). It is supplied as a high-purity solid and as DMSO stock solutions for in vitro studies.
  • Acts as an HBV core protein allosteric modulator targeting the HAP pocket.
  • Demonstrated antiviral activity (EC50 3.32 μM) in cell-based assays.
  • High purity (98.73%) and white to off-white solid appearance.
  • Chemical formula C14H16ClNO; molecular weight 249.74 g·mol⁻¹.
  • Available as solid quantities (up to 100 mg) and as a 10 mM DMSO solution.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.