Learn More
Medchemexpress LLC BA-53038B 10mg | 2306195-65-1 | 10 MG
SDP
Catalog No. 5000307625
Encompass_Preferred
BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator that binds the HAP pocket and modulates capsid assembly. It shows antiviral activity against HBV (EC50 = 3.32 μM) with low cytotoxicity (CC50 > 100 μM). Supplied as a solid research reagent with high reported purity and good solubility in DMSO.
  • Modulates HBV capsid assembly by binding the HAP pocket.
  • Antiviral activity: EC50 = 3.32 μM; low cytotoxicity (CC50 > 100 μM).
  • Solid form with reported purity of 98.73%.
  • Highly soluble in DMSO (250 mg/mL); may require ultrasonication.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (2 years); in solvent at -80°C (6 months).
Catalog No. 50-003-07625 Supplier Medchemexpress LLC Supplier No. HY11431410MG
BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator that binds the HAP pocket and modulates capsid assembly. It shows antiviral activity against HBV (EC50 = 3.32 μM) with low cytotoxicity (CC50 > 100 μM). Supplied as a solid research reagent with high reported purity and good solubility in DMSO.
  • Modulates HBV capsid assembly by binding the HAP pocket.
  • Antiviral activity: EC50 = 3.32 μM; low cytotoxicity (CC50 > 100 μM).
  • Solid form with reported purity of 98.73%.
  • Highly soluble in DMSO (250 mg/mL); may require ultrasonication.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (2 years); in solvent at -80°C (6 months).