Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BA-53038B 50mg | 2306195-65-1 | 50 MG
SDP

Catalog No. 5000308206
Encompass_Preferred

BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator that binds the HAP pocket and modulates HBV capsid assembly. It exhibits antiviral activity against HBV (reported EC50 3.32 μM) and is intended for preclinical research use.

  • High purity (reported 98.73%).
  • Molecular formula: C14H16ClNO.
  • Molecular weight: 249.74 g·mol^-1.
  • Available in multiple pack sizes for research use.
  • Storage: powder at -20°C for long-term; in solvent at -80°C for short-term.

Catalog No. 50-003-08206 Supplier Medchemexpress LLC Supplier No. HY11431450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BA-53038B is a hepatitis B virus (HBV) core protein allosteric modulator that binds the HAP pocket and modulates HBV capsid assembly. It exhibits antiviral activity against HBV (reported EC50 3.32 μM) and is intended for preclinical research use.

  • High purity (reported 98.73%).
  • Molecular formula: C14H16ClNO.
  • Molecular weight: 249.74 g·mol^-1.
  • Available in multiple pack sizes for research use.
  • Storage: powder at -20°C for long-term; in solvent at -80°C for short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.