Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BA-53038B 5mg | 2306195-65-1 | 5 MG
SDP

Catalog No. 5000229965
Encompass_Preferred

BA-53038B (CAS 2306195-65-1) is an HBV core protein allosteric modulator that binds the HAP pocket and modulates capsid assembly. It exhibits antiviral activity against hepatitis B virus (EC50 = 3.32 μM) and is supplied as a solid for research use.

  • Chemical name: N-(3-chlorophenyl)bicyclo[4.1.0]heptane-7-carboxamide.
  • Cas number: 2306195-65-1.
  • Molecular formula: C14H16ClNO.
  • Molecular weight: 249.74 g·mol⁻¹.
  • Purity: 98.7%.
  • Storage: powder at -20°C for long-term stability.
  • Available pack sizes: 5 MG and larger quantities.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-29965 Supplier Medchemexpress LLC Supplier No. HY1143145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BA-53038B (CAS 2306195-65-1) is an HBV core protein allosteric modulator that binds the HAP pocket and modulates capsid assembly. It exhibits antiviral activity against hepatitis B virus (EC50 = 3.32 μM) and is supplied as a solid for research use.

  • Chemical name: N-(3-chlorophenyl)bicyclo[4.1.0]heptane-7-carboxamide.
  • Cas number: 2306195-65-1.
  • Molecular formula: C14H16ClNO.
  • Molecular weight: 249.74 g·mol⁻¹.
  • Purity: 98.7%.
  • Storage: powder at -20°C for long-term stability.
  • Available pack sizes: 5 MG and larger quantities.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.