Learn More
Cpi International BA STANDARD
SDP
Catalog No. NC9867062
Encompass
Single-Element Standards 1000microgram/mL, Element: Barium, Source: Ba2CO3, Purity: 99.999%, Matrix: 2% HNO3
Catalog No. NC9867062 Supplier Cpi International Supplier No. S4400100041
Add to Cart
Add to Cart
Single-Element Standards 1000microgram/mL, Element: Barium, Source: Ba2CO3, Purity: 99.999%, Matrix: 2% HNO3