Learn More
Motic™ BA410E Compound LED Microscopes
Catalog No. 01258027 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Binocular
Trinocular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-258-027 Binocular
01-258-028 Trinocular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-258-027 Supplier Motic™ Supplier No. 1100100404301
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The BA410E is a professional microscope platform, ready to cover multiple applications from all biomedical research environments. A Koehler illumination is designed to support the performance of the high-level CCIS Plan Fluor and Plan Apochromat objectives.
  • Swiveling, 30° inclined Siedentopf type (Trinocular: (Light Split = 20/80))
  • Intermediate tube with rotatable analyzer and bertrand lens
  • Achromat swing-out condenser, 0.9 numerical aperature
  • Iris diaphragm
  • Coaxial coarse and fine focusing adjustment
  • Built in low position rackless stage with right hand control
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types CCIS™ EC-H Plan Achromat Objectives (PL4X, PL10X, PL40X(S), PL100X(S-oil))
Nosepiece Reversed Sextuple
Head Style Binocular
Lighting Type Kohler Illumination
Eyepiece WF 10X/22 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov