Learn More
Sklar Instruments Babcock Forceps, Sklar 16 in., Fenestrated, Grasping Forceps, Straight, Latitudinally Serrated, Stainless Steel. EA.
SDP
Catalog No. 501181450 Shop All Sklar Instruments Products
Encompass_Preferred
Babcock Forceps, Sklar. 16 in., Fenestrated, Grasping Forceps, Straight, Latitudinally Serrated, Stainless Steel. Sklar Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue. They are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. This product is straight and the overall length is 16 inches.
Catalog No. 50-118-1450 Supplier Sklar Instruments Supplier No. 610111
Babcock Forceps, Sklar. 16 in., Fenestrated, Grasping Forceps, Straight, Latitudinally Serrated, Stainless Steel. Sklar Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue. They are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. This product is straight and the overall length is 16 inches.