Learn More
World Precision Instrument Babcock Tissue Forceps, 6"
SDP
Catalog No. 502539388
Encompass
  • 6" long babcock tissue forceps
  • Stainless steel
  • Non-perforated tips
  • Used to hold delicate tissue
  • Atraumatic
Catalog No. 50-253-9388 Supplier World Precision Instrument Supplier No. 504218
  • 6" long babcock tissue forceps
  • Stainless steel
  • Non-perforated tips
  • Used to hold delicate tissue
  • Atraumatic