Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

World Precision Instrument Baby Balfour Retractor, 9.5cm Spread
SDP

Supplier:  World Precision Instrument 504197

Encompass

  • Maximum spread: 9.5cm (3.7")
  • Works well for most small animals, 60 lbs. or less
  • Stainless steel

Catalog No. 50-253-9392


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.