Learn More
Fine Science Tools Baby-Mixter Hemostat
SDP
Catalog No. NC9280128
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 15.5mm/ Alloy / Material: Stainless Steel/ Length: 14cm
Catalog No. NC9280128 Supplier Fine Science Tools Supplier No. 1301314
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 15.5mm/ Alloy / Material: Stainless Steel/ Length: 14cm