Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Baby Mixter Forceps

Catalog No. 21124792 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
5.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-124-792 5.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-792 Supplier Surgical Design™ Supplier No. FG164B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Baby Mixter forceps, 5.5 in., feature angled jaws for precise clamping and tissue manipulation in tight surgical fields and postmortem dissection.

  • Length: 5.5 in.
  • Angled Mixter (right-angle) design for excellent access around vessels, ducts, and confined anatomical structures
  • Narrow, slender jaws allow precise clamping, dissection, and retraction
  • Secure ratcheted lock maintains firm grip during procedures
  • Serrated jaws provide reliable, slip-resistant holding of tissue or instruments
  • Lightweight and well-balanced for enhanced control
  • Constructed from high-quality surgical stainless steel
  • Fully autoclavable and corrosion-resistant
  • Applications: General surgery, vascular work, delicate dissection, pediatric procedures, and autopsy/postmortem tissue isolation and specimen retrieval

Specifications

Product Type Forceps
Material Stainless Steel
Length (English) 5.5 in.
Length (Metric) 14 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.