Learn More
Cayman Chemical YM-254890, Purity: ≥95%, CAS: 568580-02-9, 1 mg
SDP
Catalog No. 502988833
Encompass_Preferred
A cyclic depsipeptide Gαq/11 inhibitor; inhibits Gαq/11-mediated intracellular calcium mobilization induced by 2MeSADP or methacholine in C6-15 cells expressing the purinergic P2Y1 receptor (IC50 = 0.15 µM) and in CHO cells expressing M1 muscarinic acetylcholine receptors (IC50 = 0.15 µM), respectively; inhibits ADP-, collagen-, TRAP-, arachidonic acid-, or U46619-induced platelet aggregation in isolated human platelet-rich plasma (IC50s = 0.39, 0.15, 0.71, 0.25, and 0.34 µM, respectively); reduces shear stress-induced thrombus formation in isolated human whole blood; reduces platelet thrombus formation in a cynomolgus monkey model of femoral artery thrombosis at 1, 3, and 10 µg/kg
Catalog No. 50-298-8833 Supplier Cayman Chemical Supplier No. 297351 MG
A cyclic depsipeptide Gαq/11 inhibitor; inhibits Gαq/11-mediated intracellular calcium mobilization induced by 2MeSADP or methacholine in C6-15 cells expressing the purinergic P2Y1 receptor (IC50 = 0.15 µM) and in CHO cells expressing M1 muscarinic acetylcholine receptors (IC50 = 0.15 µM), respectively; inhibits ADP-, collagen-, TRAP-, arachidonic acid-, or U46619-induced platelet aggregation in isolated human platelet-rich plasma (IC50s = 0.39, 0.15, 0.71, 0.25, and 0.34 µM, respectively); reduces shear stress-induced thrombus formation in isolated human whole blood; reduces platelet thrombus formation in a cynomolgus monkey model of femoral artery thrombosis at 1, 3, and 10 µg/kg