Learn More
Bioworld BAC DNA MiniPrep Kit, 25 Prep(s)
Catalog No. 50196958
Encompass_Preferred
For the rapid preparation of BAC DNA High DNA recovery Alcohol-free protocol Convenient spin column protocol FEATURES AND BENEFITSHigh Recovery -0.6 to 1 µg of BAC DNA can be purified from 3.0 mL of bacterial culture. Alcohol-free protocol -No alcohols are used durig the purification process, thus the DNA is compatible with alcohol sensitive downstream applications. Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 1 hour. Recovered DNA is suitable for downstream applications - Purified BAC DNA is fully compatible with restriction enzyme digestions and sequencing. High binding capacity of columns -The binding capacity of the columns in the BAC DNA MiniPrep Kit is 25 µg
Catalog No. 50-196-958 Supplier Bioworld Supplier No. 107601011
For the rapid preparation of BAC DNA High DNA recovery Alcohol-free protocol Convenient spin column protocol FEATURES AND BENEFITSHigh Recovery -0.6 to 1 µg of BAC DNA can be purified from 3.0 mL of bacterial culture. Alcohol-free protocol -No alcohols are used durig the purification process, thus the DNA is compatible with alcohol sensitive downstream applications. Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 1 hour. Recovered DNA is suitable for downstream applications - Purified BAC DNA is fully compatible with restriction enzyme digestions and sequencing. High binding capacity of columns -The binding capacity of the columns in the BAC DNA MiniPrep Kit is 25 µg