Learn More
Cayman Chemical IB-96212, Purity: ≥70%, CAS: 220858-11-7, 25 mg
SDP
Catalog No. 502944206
Encompass_Preferred
A macrolide with antibacterial and anticancer activities; active against M. luteus, B. subtilis, and S. aureus (MICs = 0.4, 100, and 100 µg/ml, respectively); induces death in P388, A549, HT-29, and SK-MEL-28 cancer cells (IC50s = 0.0001, 1, 1, and 1 µg/ml, respectively)
Catalog No. 50-294-4206 Supplier Cayman Chemical Supplier No. 4093825 MG
A macrolide with antibacterial and anticancer activities; active against M. luteus, B. subtilis, and S. aureus (MICs = 0.4, 100, and 100 µg/ml, respectively); induces death in P388, A549, HT-29, and SK-MEL-28 cancer cells (IC50s = 0.0001, 1, 1, and 1 µg/ml, respectively)