Learn More
Takara Bio BacPAK™ Baculovirus Rapid Titer Kit, 5 Assays
SDP
Catalog No. NC9445196
Encompass
Kit for quick determination of the titer of baculovirus stocks (e.g. stocks of recombinant baculovirus). A titer is obtained in 2 days, compared to 5–8 days with conventional titering methods. The assay uses an antibody to detect virus infected cells, and is thus able to detect virus infections long before methods that detect plaque formation. Stocks of any AcMNPV-type baculovirus with titers of >104PFU/ml or IFU/ml can be titered using this kit. 5 Assays
Catalog No. NC9445196 Supplier Takara Bio Supplier No. 631406
Kit for quick determination of the titer of baculovirus stocks (e.g. stocks of recombinant baculovirus). A titer is obtained in 2 days, compared to 5–8 days with conventional titering methods. The assay uses an antibody to detect virus infected cells, and is thus able to detect virus infections long before methods that detect plaque formation. Stocks of any AcMNPV-type baculovirus with titers of >104PFU/ml or IFU/ml can be titered using this kit. 5 Assays