Learn More
Medchemexpress LLC Bacampicillin (hydrochloride) | 37661-08-8 | 99.9% | 1 ML
SDP
Catalog No. 5000411329
Encompass_Preferred
Bacampicillin hydrochloride is a penicillin antibiotic that serves as a prodrug of ampicillin. It is notable for its improved oral bioavailability compared to ampicillin.
  • It is a penicillin antibiotic.
  • It acts as a prodrug of ampicillin.
  • It offers improved oral bioavailability.
  • It is intended for research use only.
Catalog No. 50-004-11329 Supplier Medchemexpress LLC Supplier No. HYB1149A10MM/1ML
Bacampicillin hydrochloride is a penicillin antibiotic that serves as a prodrug of ampicillin. It is notable for its improved oral bioavailability compared to ampicillin.
  • It is a penicillin antibiotic.
  • It acts as a prodrug of ampicillin.
  • It offers improved oral bioavailability.
  • It is intended for research use only.