Learn More
Medchemexpress LLC Bacampicillin (hydrochloride) | 37661-08-8 | 99.9% | 50 MG
SDP
Catalog No. 5000413343
Encompass_Preferred
Bacampicillin hydrochloride is a penicillin antibiotic and a prodrug of ampicillin with improved oral bioavailability. It acts as an inhibitor targeting β-lactam.
  • Penicillin antibiotic
  • Prodrug of ampicillin
  • Improved oral bioavailability
  • Target: β-lactam
  • Applicable to bacterial infection research
Catalog No. 50-004-13343 Supplier Medchemexpress LLC Supplier No. HYB1149A50MG
Bacampicillin hydrochloride is a penicillin antibiotic and a prodrug of ampicillin with improved oral bioavailability. It acts as an inhibitor targeting β-lactam.
  • Penicillin antibiotic
  • Prodrug of ampicillin
  • Improved oral bioavailability
  • Target: β-lactam
  • Applicable to bacterial infection research