Learn More
Cayman Chemical BaccatIn III 10mg
SDP
Catalog No. 502995394
Encompass_Preferred
A precursor of paclitaxel and an apoptotic agent; induces apoptosis in JR4-Jurkat leukemia, HepG2 liver hepatocellular carcinoma, HeLa cervical, OVCAR-3 ovarian carcinoma, and T47D breast cancer cell lines (IC50s = 3.5, 3, 4, 5, and 2 μM, respectively); increases APC capacity of BM-DCs; decreases tumor growth of 4T1 mammary carcinoma and CT26 colon carcinoma flank implants as well as inhibits the accumulation and activity of MDSCs in spleen in immunocompetent mice
Catalog No. 50-299-5394 Supplier Cayman Chemical Supplier No. 2140410 MG
A precursor of paclitaxel and an apoptotic agent; induces apoptosis in JR4-Jurkat leukemia, HepG2 liver hepatocellular carcinoma, HeLa cervical, OVCAR-3 ovarian carcinoma, and T47D breast cancer cell lines (IC50s = 3.5, 3, 4, 5, and 2 μM, respectively); increases APC capacity of BM-DCs; decreases tumor growth of 4T1 mammary carcinoma and CT26 colon carcinoma flank implants as well as inhibits the accumulation and activity of MDSCs in spleen in immunocompetent mice