Learn More
TARGETMOL CHEMICALS INC BACCATIN III 25MG
SDP
Catalog No. 502577047
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baccatin III (Baccatin Ⅲ) is a polycyclic diterpene which can be used for the semi-synthesis of paclitaxel and analogs. It has immunomodulatory and anticancer chemotherapeutic activities. purity: 98%
Catalog No. 50-257-7047 Supplier TARGETMOL CHEMICALS INC Supplier No. T278825MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baccatin III (Baccatin Ⅲ) is a polycyclic diterpene which can be used for the semi-synthesis of paclitaxel and analogs. It has immunomodulatory and anticancer chemotherapeutic activities. purity: 98%