Learn More
Medchemexpress LLC Baccatin III | 27548-93-2 | 1 ML
SDP
Catalog No. 5000419158
Encompass_Preferred
Baccatin III is an orally available, selective inhibitor of the TGF-β1 signaling pathway and myeloid-derived suppressor cell (MDSC) activation. It targets the AKT/STAT6 and Smad2/3 pathways, blocking TGF-β1-induced fibroblast differentiation and MDSC-mediated immunosuppression.
  • Exerts anti-inflammatory and anti-fibrotic effects by inhibiting macrophage activation and extracellular matrix deposition.
  • Shows potential in the treatment of pulmonary fibrosis and cancer by regulating the tumor immune microenvironment.
Catalog No. 50-004-19158 Supplier Medchemexpress LLC Supplier No. HYN698510MM/1ML
Baccatin III is an orally available, selective inhibitor of the TGF-β1 signaling pathway and myeloid-derived suppressor cell (MDSC) activation. It targets the AKT/STAT6 and Smad2/3 pathways, blocking TGF-β1-induced fibroblast differentiation and MDSC-mediated immunosuppression.
  • Exerts anti-inflammatory and anti-fibrotic effects by inhibiting macrophage activation and extracellular matrix deposition.
  • Shows potential in the treatment of pulmonary fibrosis and cancer by regulating the tumor immune microenvironment.