Learn More
Cayman Chemical Baccatin III, Purity: ≥98%, CAS: 27548-93-2, 5 mg
SDP
Catalog No. 502994082
Encompass_Preferred
A precursor of paclitaxel and an apoptotic agent; induces apoptosis in JR4-Jurkat leukemia, HepG2 liver hepatocellular carcinoma, HeLa cervical, OVCAR-3 ovarian carcinoma, and T47D breast cancer cell lines (IC50s = 3.5, 3, 4, 5, and 2 μM, respectively); increases APC capacity of BM-DCs; decreases tumor growth of 4T1 mammary carcinoma and CT26 colon carcinoma flank implants as well as inhibits the accumulation and activity of MDSCs in spleen in immunocompetent mice
Catalog No. 50-299-4082 Supplier Cayman Chemical Supplier No. 214045 MG
A precursor of paclitaxel and an apoptotic agent; induces apoptosis in JR4-Jurkat leukemia, HepG2 liver hepatocellular carcinoma, HeLa cervical, OVCAR-3 ovarian carcinoma, and T47D breast cancer cell lines (IC50s = 3.5, 3, 4, 5, and 2 μM, respectively); increases APC capacity of BM-DCs; decreases tumor growth of 4T1 mammary carcinoma and CT26 colon carcinoma flank implants as well as inhibits the accumulation and activity of MDSCs in spleen in immunocompetent mice