Learn More
Medchemexpress LLC HY-N6985 5mg Medchemexpress, Baccatin III CAS:27548-93-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69855MG
Encompass_Preferred
Medchemexpress, HY-N6985 5mg Baccatin III CAS:27548-93-2 Baccatin III is a natural product isolated from Pacific yew tree and related species. Baccatin III reduces tumor progression by inhibiting the accumulation and suppressive function of MDSCs [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3975