Learn More
ACROBiosystems Recombinant Protein;Human BACE-1 Protein, Fc Tag;HEK293;1MG;BA1,BACE1,BACE-1,ASP2,BACE,FLJ90568,HSPC104,KIAA1149,memapsin-2,memapsin 2
Catalog No. 501659027
Encompass_Preferred
Recombinant Protein;Human BACE-1 Protein, Fc Tag;1MG;Human BACE-1, Fc Tag (BA1-H5261) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;Human BACE-1, Fc Tag is fused with a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 75 kDa. The predicted N-terminus is Glu. The reducing (R) protein migrates as 100-110 kDa in SDS-PAGE due to glycosylation.
Catalog No. 50-165-9027 Supplier ACROBiosystems Supplier No. BA1H52611MG
Recombinant Protein;Human BACE-1 Protein, Fc Tag;1MG;Human BACE-1, Fc Tag (BA1-H5261) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;Human BACE-1, Fc Tag is fused with a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 75 kDa. The predicted N-terminus is Glu. The reducing (R) protein migrates as 100-110 kDa in SDS-PAGE due to glycosylation.