Learn More
ACROBiosystems Recombinant Protein;Human BACE-1 Protein;HEK293;1MG;BA1,BACE1,BACE-1,ASP2,BACE,FLJ90568,HSPC104,KIAA1149,memapsin-2,memapsin 2
Catalog No. 501659029
Encompass_Preferred
Recombinant Protein;Human BACE-1 Protein;1MG;Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9029 Supplier ACROBiosystems Supplier No. BA1H52201MG
Recombinant Protein;Human BACE-1 Protein;1MG;Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.