Learn More
Aviva Systems Biology Corp BACE2 Antibody - N-terminal region (ARP46853_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46853_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BACE2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BACE2. Alternate names: AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAPApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54021